Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-methoxynicotinic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

26180-1G 26180-25G 26180-500MG 26180-5G
76900-284EA 350.1 USD InStock
76900-284 76900-286 76900-288 76900-290
2-Fluoro-4-methoxynicotinic acid ≥97% research grade
2-Fluoro-4-methoxynicotinic acid


Formula: C₇H₆FNO₃
MW: 171.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12828253
CAS Number: 1190315-81-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty