Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Difluoro-6-bromophenylboronic acid pinacol ester ≥95% research grade

Supplier: AOB CHEM USA
Warning

26500-1G 26500-25G 26500-500MG 26500-5G
76900-378EA 326.76 USD InStock
76900-378 76900-380 76900-382 76900-384
2,4-Difluoro-6-bromophenylboronic acid pinacol ester ≥95% research grade
2,4-Difluoro-6-bromophenylboronic acid pinacol ester


Formula: C₁₂H₁₄BBrF₂O₂
MW: 318.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977227
CAS Number: 2121513-38-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty