Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-(pyrrolidin-1-yl)benzaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

26767-1G 26767-25G 26767-500MG 26767-5G
76900-454EA 155.96 USD InStock
76900-454 76900-456 76900-458 76900-460
2-Bromo-6-(pyrrolidin-1-yl)benzaldehyde ≥97% research grade
2-Bromo-6-(pyrrolidin-1-yl)benzaldehyde


Formula: C₁₁H₁₂BrNO
Storage Temperature: Refrigerator
MDL Number: MFCD22205829
CAS Number: 1375069-38-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty