Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-(chloromethyl)-3-methylpyridine ≥97% research grade

Supplier: AOB CHEM USA
Warning

26773-1G 26773-25G 26773-500MG 26773-5G
76900-468EA 264.17 USD InStock
76900-468 76900-470 76900-472 76900-474
2-Chloro-5-(chloromethyl)-3-methylpyridine ≥97% research grade
2-Chloro-5-(chloromethyl)-3-methylpyridine


Formula: C₇H₇Cl₂N
MW: 176.043 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09261084
CAS Number: 150807-88-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty