Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2,4-difluorobenzeneboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

26780-25G 26780-5G
76900-482EA 453.01 USD InStock
76900-482 76900-484
5-Bromo-2,4-difluorobenzeneboronic acid ≥97%
5-Bromo-2,4-difluorobenzeneboronic acid


Formula: C₆H₄BBrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18783127
CAS Number: 2096331-76-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty