Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Hydroxy-4-methylphenyl)boronic acid 95% research grade

Supplier: AOB CHEM USA
Warning

27324-1G 27324-500MG 27324-5G 27324-25G
76900-658EA 558.04 USD InStock
76900-658 76900-660 76900-662 77654-616
(2-Hydroxy-4-methylphenyl)boronic acid 95% research grade
(2-Hydroxy-4-methylphenyl)boronic acid


Formula: C₇H₉BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD18397304
CAS Number: 259209-25-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty