Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Difluoro-5-methyl-boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

27954-1G 27954-25G 27954-5G 27954-500MG
76900-850EA 66.84 USD InStock
76900-850 76900-852 76900-854 77645-682
2,4-Difluoro-5-methyl-boronic acid ≥97% research grade
2,4-Difluoro-5-methyl-boronic acid


Formula: C₇H₇BF₂O₂
MW: 171.937 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10697417
CAS Number: 900175-09-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty