Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Isopropoxypyridine-4-boronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

28359-1G 28359-25G 28359-500MG 28359-5G
76900-966EA 379.81 USD InStock
76900-966 76900-968 76900-970 76900-972
2-Isopropoxypyridine-4-boronic acid pinacol ester ≥97%
2-Isopropoxypyridine-4-boronic acid pinacol ester


Formula: C₁₄H₂₂BNO₃
Storage Temperature: Ambient
MDL Number: MFCD11878168
CAS Number: 1257554-10-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty