Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-3-methylphenylboronic acid pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

28720-1G 28720-25G 28720-500MG 28720-5G
76901-048EA 126.25 USD InStock
76901-048 76901-050 76901-052 76901-054
2-Hydroxy-3-methylphenylboronic acid pinacol ester ≥97% research grade
2-Hydroxy-3-methylphenylboronic acid pinacol ester


Formula: C₁₃H₁₉BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16994284
CAS Number: 2095797-27-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty