Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-iodobenzyl alcohol ≥97%

Supplier: AOB CHEM USA
Warning

28963-1G 28963-25G 28963-500MG 28963-5G
76901-090EA 364.95 USD InStock
76901-090 76901-092 76901-094 76901-096
2-Chloro-5-iodobenzyl alcohol ≥97%
2-Chloro-5-iodobenzyl alcohol
Precaución: for Research Use Only


Formula: C₇H₆ClIO
MW: 268.479 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18072886
CAS Number: 1035155-69-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty