Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzyloxy-4-(trifluoromethoxy)phenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

29194-1G 29194-25G 29194-500MG 29194-5G
76901-152EA 253.56 USD InStock
76901-152 76901-154 76901-156 76901-158
2-Benzyloxy-4-(trifluoromethoxy)phenylboronic acid ≥97% research grade
2-Benzyloxy-4-(trifluoromethoxy)phenylboronic acid


Formula: C₁₄H₁₂BF₃O₄
Storage Temperature: Refrigerator
MDL Number: MFCD28384207
CAS Number: 1704064-19-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty