Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Difluoro-6-methylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

29245-1G 29245-25G 29245-500MG 29245-5G
76901-168EA 245.07 USD InStock
76901-168 76901-170 76901-172 76901-174
2,5-Difluoro-6-methylphenylboronic acid ≥97%
2,5-Difluoro-6-methylphenylboronic acid


Formula: C₇H₇BF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD23712898
CAS Number: 1884277-68-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty