Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-chloro-3-iodopyridine ≥95%

Supplier: AOB CHEM USA
Warning

30419-1G 30419-25G 30419-500MG 30419-5G
76901-520EA 290.69 USD InStock
76901-520 76901-522 76901-524 76901-526
2-Bromo-5-chloro-3-iodopyridine ≥95%
2-Bromo-5-chloro-3-iodopyridine


Formula: C₅H₂BrClIN
Storage Temperature: Refrigerator
MDL Number: MFCD13185353
CAS Number: 1211535-24-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty