Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-6-methoxybenzeneboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

30429-25G 30429-5G
76901-536EA 524.09 USD InStock
76901-536 76901-538
2-Fluoro-6-methoxybenzeneboronic acid pinacol ester ≥97%
2-Fluoro-6-methoxybenzeneboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₈BFO₃
MW: 252.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16996294
CAS Number: 1599432-41-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty