Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Difluoro-6-iodophenol ≥97%

Supplier: AOB CHEM USA
Warning

30444-1G 30444-25G 30444-500MG 30444-5G
76901-552EA 177.18 USD InStock
76901-552 76901-554 76901-556 76901-558
2,4-Difluoro-6-iodophenol ≥97%
2,4-Difluoro-6-iodophenol


Formula: C₆H₃F₂IO
Storage Temperature: Ambient
MDL Number: MFCD00466295
CAS Number: 551002-42-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty