Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-(tert-butyl)benzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

30639-1G 30639-25G 30639-500MG 30639-5G
76901-658EA 162.32 USD InStock
76901-658 76901-660 76901-662 76901-664
2-Bromo-4-(tert-butyl)benzoic acid ≥97% research grade
2-Bromo-4-(tert-butyl)benzoic acid


Formula: C₁₁H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD11040238
CAS Number: 6332-96-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty