Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethylphenylboronic acid pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

31110-1G 31110-25G 31110-500MG 31110-5G
76901-892EA 300.24 USD InStock
76901-892 76901-894 76901-896 76901-898
2-Ethylphenylboronic acid pinacol ester ≥97% research grade
2-Ethylphenylboronic acid pinacol ester


Formula: C₁₄H₂₁BO₂
MW: 232.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18779859
CAS Number: 1683582-61-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty