Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylbiphenyl-4-ylboronic acid pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

31290-1G 31290-500MG 31290-5G
76901-942EA 594.11 USD InStock
76901-942 76901-944 76901-946
2-Methylbiphenyl-4-ylboronic acid pinacol ester ≥97% research grade
2-Methylbiphenyl-4-ylboronic acid pinacol ester


Formula: C₁₉H₂₃BO₂
Storage Temperature: Ambient
MDL Number: MFCD31806012
CAS Number: 2649399-93-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty