Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-3-(methoxycarbonyl)-5-pyridineboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

31899-1G 31899-25G 31899-500MG 31899-5G
76902-108EA 242.95 USD InStock
76902-108 76902-110 76902-112 76902-114
2-Methoxy-3-(methoxycarbonyl)-5-pyridineboronic acid pinacol ester ≥97%
2-Methoxy-3-(methoxycarbonyl)-5-pyridineboronic acid pinacol ester


Formula: C₁₄H₂₀BNO₅
Storage Temperature: Ambient
MDL Number: MFCD11855973
CAS Number: 1083168-93-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty