Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyano-4-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

31992-25G 31992-5G
76902-150EA 456.19 USD InStock
76902-150 76902-152
2-Cyano-4-fluorophenylboronic acid pinacol ester ≥97%
2-Cyano-4-fluorophenylboronic acid pinacol ester


Formula: C₁₃H₁₅BFNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09260444
CAS Number: 461451-63-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty