Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-iodo-3-methoxypyridine ≥97% research grade

Supplier: AOB CHEM USA
Warning

32088-1G 32088-25G 32088-500MG 32088-5G
76902-180EA 144.29 USD InStock
76902-180 76902-182 76902-184 76902-186
2-Chloro-4-iodo-3-methoxypyridine ≥97% research grade
2-Chloro-4-iodo-3-methoxypyridine


Formula: C₆H₅ClINO
Storage Temperature: Refrigerator
MDL Number: MFCD16610523
CAS Number: 1227592-88-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty