Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-(Methylsulfanyl)Benzaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

33285-1G 33285-25G 33285-500MG 33285-5G
76902-530EA 188.85 USD InStock
76902-530 76902-532 76902-534 76902-536
2-Chloro-6-(Methylsulfanyl)Benzaldehyde ≥97% research grade
2-Chloro-6-(Methylsulfanyl)Benzaldehyde


Formula: C₈H₇ClOS
Storage Temperature: Ambient
MDL Number: MFCD18887987
CAS Number: 201987-39-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty