Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Difluoro-4-isopropoxyphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

33345-1G 33345-25G 33345-5G
76902-546EA 112.46 USD InStock
76902-546 76902-548 76902-550
2,5-Difluoro-4-isopropoxyphenylboronic acid ≥97%
2,5-Difluoro-4-isopropoxyphenylboronic acid


Formula: C₉H₁₁BF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD30834049
CAS Number: 2121515-24-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty