Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dimethyl-3-fluorobenzyl alcohol ≥97%

Supplier: AOB CHEM USA
Warning

33642-1G 33642-25G 33642-500MG 33642-5G
76902-660EA 242.95 USD InStock
76902-660 76902-662 76902-664 76902-666
2,4-Dimethyl-3-fluorobenzyl alcohol ≥97%
2,4-Dimethyl-3-fluorobenzyl alcohol


Formula: C₉H₁₁FO
Storage Temperature: Refrigerator
MDL Number: MFCD07777169
CAS Number: 26583-82-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty