Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-3-isopropylbenzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

34273-1G 34273-25G 34273-500MG 34273-5G
76902-838EA 206.88 USD InStock
76902-838 76902-840 76902-842 76902-844
2-Hydroxy-3-isopropylbenzaldehyde ≥97%
2-Hydroxy-3-isopropylbenzaldehyde
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11040457
CAS Number: 67372-96-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty