Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-3-hydroxybenzonitrile ≥97%

Supplier: AOB CHEM USA
Warning

34850-1G 34850-25G 34850-500MG 34850-5G
76903-032EA 290.69 USD InStock
76903-032 76903-034 76903-036 76903-038
2,6-Dichloro-3-hydroxybenzonitrile ≥97%
2,6-Dichloro-3-hydroxybenzonitrile
Precaución: for Research Use Only


Formula: C₇H₃Cl₂NO
MW: 188.011 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01662155
CAS Number: 3336-34-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty