Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(3-Bromophenyl)thiophene ≥97%

Supplier: AOB CHEM USA
Warning

35522-1G 35522-25G 35522-500MG 35522-5G
76903-132EA 264.17 USD InStock
76903-132 76903-134 76903-136 76903-138
3-(3-Bromophenyl)thiophene ≥97%
3-(3-Bromophenyl)thiophene
Precaución: for Research Use Only


Formula: C₁₀H₇BrS
Storage Temperature: Refrigerator
MDL Number: MFCD06740296
CAS Number: 20608-84-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty