Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4,5-dichlorobenzenamine ≥97%

Supplier: AOB CHEM USA
Warning

35896-1G 35896-25G 35896-500MG 35896-5G
76903-228EA 183.54 USD InStock
76903-228 76903-230 76903-232 76903-234
2-Bromo-4,5-dichlorobenzenamine ≥97%
2-Bromo-4,5-dichlorobenzenamine
Precaución: for Research Use Only


Formula: C₆H₄BrCl₂N
Storage Temperature: Refrigerator
MDL Number: MFCD00672938
CAS Number: 1940-30-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty