Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Difluoro-3-iodobenzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

36358-1G 36358-25G 36358-500MG 36358-5G
76903-254EA 187.78 USD InStock
76903-254 76903-256 76903-258 76903-260
2,4-Difluoro-3-iodobenzaldehyde ≥97%
2,4-Difluoro-3-iodobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₃F₂IO
Storage Temperature: Refrigerator
MDL Number: MFCD28789180
CAS Number: 1806292-43-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty