Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-(methoxymethoxy)-4-(trifluoromethyl)benzaldehyde ≥95%

Supplier: AOB CHEM USA
Warning

37231-1G 37231-25G 37231-500MG 37231-5G
76903-448EA 209 USD InStock
76903-448 76903-450 76903-452 76903-454
2-Fluoro-3-(methoxymethoxy)-4-(trifluoromethyl)benzaldehyde ≥95%
2-Fluoro-3-(methoxymethoxy)-4-(trifluoromethyl)benzaldehyde
Precaución: for Research Use Only


Formula: C₁₀H₈F₄O₃
MW: 252.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632169
CAS Number: 2379322-61-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty