Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Butoxy-6-fluorobenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

38579-1G 38579-25G 38579-500MG 38579-5G
76903-682EA 260.99 USD InStock
76903-682 76903-684 76903-686 76903-688
2-Butoxy-6-fluorobenzoic acid ≥97%
2-Butoxy-6-fluorobenzoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₃FO₃
Storage Temperature: Refrigerator
MDL Number: MFCD21944929
CAS Number: 1499793-21-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty