Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-methoxy-5-methylphenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

40886-1G 40886-25G 40886-500MG 40886-5G
76903-838EA 162.32 USD InStock
76903-838 76903-840 76903-842 76903-844
2-Fluoro-3-methoxy-5-methylphenylboronic acid ≥97% research grade
2-Fluoro-3-methoxy-5-methylphenylboronic acid


Formula: C₈H₁₀BFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD19440599
CAS Number: 2096329-52-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty