Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoro-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97%

Supplier: AOB CHEM USA
Warning

43634-1G 43634-25G 43634-500MG 43634-5G
76903-930EA 217.49 USD InStock
76903-930 76903-932 76903-934 76903-936
2,6-Difluoro-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97%
2,6-Difluoro-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Precaución: for Research Use Only


Formula: C₁₂H₁₅BF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD24039815
CAS Number: 2756163-62-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty