Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Formyl-4-methylthiophene-3-carboxylic acid ≥95%

Supplier: AOB CHEM USA
Warning

45934-1G 45934-25G 45934-500MG 45934-5G
76904-020EA 334.19 USD InStock
76904-020 76904-022 76904-024 76904-026
2-Formyl-4-methylthiophene-3-carboxylic acid ≥95%
2-Formyl-4-methylthiophene-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₇H₆O₃S
Storage Temperature: Refrigerator
MDL Number: MFCD32899458
CAS Number: 2167543-72-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty