Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-(trifluoromethoxy)benzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

47532-1G 47532-500MG 47532-5G
76904-166EA 468.92 USD InStock
76904-166 76904-168 76904-170
2-Chloro-4-(trifluoromethoxy)benzoic acid ≥97% research grade
2-Chloro-4-(trifluoromethoxy)benzoic acid


Formula: C₈H₄ClF₃O₃
MW: 240.564 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18393797
CAS Number: 1261731-19-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty