Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethoxy-4-fluorobenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

48384-1G 48384-25G 48384-500MG 48384-5G
76904-280EA 206.88 USD InStock
76904-280 76904-282 76904-284 76904-286
2-Ethoxy-4-fluorobenzoic acid ≥97% research grade
2-Ethoxy-4-fluorobenzoic acid


Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD14582790
CAS Number: 1233541-55-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty