Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-phenoxybenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

49658-1G 49658-500MG 49658-5G
76904-530EA 589.87 USD InStock
76904-530 76904-532 76904-534
2-Chloro-4-phenoxybenzoic acid ≥97%
2-Chloro-4-phenoxybenzoic acid
Precaución: for Research Use Only


Formula: C₁₃H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD18044308
CAS Number: 860697-53-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty