Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-methoxy-4-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

50194-1G 50194-25G 50194-500MG 50194-5G
76904-570EA 441.34 USD InStock
76904-570 76904-572 76904-574 76904-576
2-Fluoro-3-methoxy-4-(trifluoromethyl)benzoic acid ≥97%
2-Fluoro-3-methoxy-4-(trifluoromethyl)benzoic acid
Precaución: for Research Use Only


Formula: C₉H₆F₄O₃
MW: 238.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902871
CAS Number: 2387102-13-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty