Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3,5-dimethylphenol ≥97%

Supplier: AOB CHEM USA
Warning

50619-1G 50619-25G 50619-500MG 50619-5G
76904-586EA 381.93 USD InStock
76904-586 76904-588 76904-590 76904-592
2-Chloro-3,5-dimethylphenol ≥97%
2-Chloro-3,5-dimethylphenol


Formula: C₈H₉ClO
Storage Temperature: Ambient
MDL Number: MFCD09835163
CAS Number: 5538-41-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty