Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethyl-4-methoxybenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

51203-1G 51203-25G 51203-500MG 51203-5G
76904-630EA 439.22 USD InStock
76904-630 76904-632 76904-634 76904-636
2-Ethyl-4-methoxybenzoic acid ≥97%
2-Ethyl-4-methoxybenzoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD18411999
CAS Number: 103264-62-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty