Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-[1,1'-biphenyl]-4-carboxylic acid ≥97%

Supplier: AOB CHEM USA
Warning

51573-1G 51573-25G 51573-5G 51573-500MG
76904-686EA 117.76 USD InStock
76904-686 76904-688 76904-690 77647-486
2-Methyl-[1,1'-biphenyl]-4-carboxylic acid ≥97%
2-Methyl-[1,1'-biphenyl]-4-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11554176
CAS Number: 178313-67-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty