Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3-(difluoromethoxy)pyridine ≥97% research grade

Supplier: AOB CHEM USA
Warning

51799-1G 51799-25G 51799-500MG 51799-5G
76904-698EA 144.29 USD InStock
76904-698 76904-700 76904-702 76904-704
2-Chloro-3-(difluoromethoxy)pyridine ≥97% research grade
2-Chloro-3-(difluoromethoxy)pyridine


Formula: C₆H₄ClF₂NO
MW: 179.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185847
CAS Number: 1206977-80-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty