Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-(methylthio)pyridine ≥95% research grade

Supplier: AOB CHEM USA
Warning

52755-1G 52755-25G 52755-5G
76904-852EA 88.06 USD InStock
76904-852 76904-854 76904-856
2-Bromo-6-(methylthio)pyridine ≥95% research grade
2-Bromo-6-(methylthio)pyridine


Formula: C₆H₆BrNS
Storage Temperature: Ambient
MDL Number: MFCD09265530
CAS Number: 74134-42-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty