Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Trifluoromethoxy)salicylaldehyde ≥95%

Supplier: AOB CHEM USA
Warning

54865-1G 54865-25G 54865-500MG 54865-5G
76905-096EA 144.29 USD InStock
76905-096 76905-098 76905-100 76905-102
3-(Trifluoromethoxy)salicylaldehyde ≥95%
3-(Trifluoromethoxy)salicylaldehyde
Precaución: for Research Use Only


Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03788527
CAS Number: 497959-31-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty