Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dimethoxysalicylaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

60089-1G 60089-25G 60089-500MG 60089-5G
76905-356EA 257.8 USD InStock
76905-356 76905-358 76905-360 76905-362
3,4-Dimethoxysalicylaldehyde ≥97%
3,4-Dimethoxysalicylaldehyde
Precaución: for Research Use Only


Formula: C₉H₁₀O₄
MW: 182.1733 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016588
CAS Number: 19283-70-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty