Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-1,3,5-trifluoro-4-iodobenzene ≥97%

Supplier: AOB CHEM USA
Warning

61105-1G 61105-25G 61105-500MG 61105-5G
76905-446EA 134.74 USD InStock
76905-446 76905-448 76905-450 76905-452
2-Bromo-1,3,5-trifluoro-4-iodobenzene ≥97%
2-Bromo-1,3,5-trifluoro-4-iodobenzene
Precaución: for Research Use Only


Formula: C₆HBrF₃I
Storage Temperature: Refrigerator
CAS Number: 2764729-86-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty