Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-(1-piperidinyl)pyridine ≥97% research grade

Supplier: AOB CHEM USA
Warning

61167-1G 61167-25G 61167-500MG 61167-5G
76905-458EA 144.29 USD InStock
76905-458 76905-460 76905-462 76905-464
2-Chloro-6-(1-piperidinyl)pyridine ≥97% research grade
2-Chloro-6-(1-piperidinyl)pyridine


Formula: C₁₀H₁₃ClN₂
MW: 196.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13561951
CAS Number: 19946-28-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty