Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichloro-4-fluorobenzoic acid ≥95%

Supplier: AOB CHEM USA
Warning

61662-1G 61662-25G 61662-500MG 61662-5G
76905-474EA 436.03 USD InStock
76905-474 76905-476 76905-478 76905-480
2,5-Dichloro-4-fluorobenzoic acid ≥95%
2,5-Dichloro-4-fluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃Cl₂FO₂
MW: 209.0019 g/mol
Storage Temperature: Ambient
CAS Number: 35989-28-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty