Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-ethoxybenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

63224-1G 63224-25G 63224-500MG 63224-5G
76905-596EA 153.84 USD InStock
76905-596 76905-598 76905-600 76905-602
2-Chloro-6-ethoxybenzoic acid ≥97% research grade
2-Chloro-6-ethoxybenzoic acid


Formula: C₉H₉ClO₃
MW: 200.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11156593
CAS Number: 1020998-62-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty