Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoro-3,5-dimethoxybenzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

63706-1G 63706-25G 63706-500MG 63706-5G
76905-640EA 159.14 USD InStock
76905-640 76905-642 76905-644 76905-646
2,6-Difluoro-3,5-dimethoxybenzaldehyde ≥97%
2,6-Difluoro-3,5-dimethoxybenzaldehyde


Formula: C₉H₈F₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD29058863
CAS Number: 1890954-24-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty